LIF388583860 Life Chemicals Inc.

(4-chlorobenzyl)hydrazine hydrochloride

  • MDL number MFCD01722572
  • Cas Number : 75333-04-9
  • Molecular Weight : 193.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart