LIF388583862 Life Chemicals Inc.

1-(Piperidin-4-ylmethyl)-1H-benzimidazole dihydrochloride

  • MDL number MFCD16661024
  • Cas Number : 1351589-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart