LIF388583866 Life Chemicals Inc.

[4-(morpholin-4-ylmethyl)phenyl]boronic acid hydrochloride

  • MDL number MFCD16661158
  • Cas Number : 1335234-36-0
  • Molecular Weight : 257.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart