LIF388583869 Life Chemicals Inc.

[(3-bromophenyl)methyl]hydrazine hydrochloride

  • MDL number MFCD16660424
  • Cas Number : 1384427-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart