LIF388583870 Life Chemicals Inc.

(3,5-dimethoxybenzyl)hydrazine hydrochloride

  • MDL number MFCD16660423
  • Cas Number : 1351648-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart