LIF388583871 Life Chemicals Inc.

(2,5-dimethylbenzyl)hydrazine hydrochloride

  • MDL number MFCD16660418
  • Cas Number : 1351630-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart