LIF388583872 Life Chemicals Inc.

[(3-pyrimidin-2-yl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride

  • MDL number MFCD16661140
  • Cas Number : 1351646-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart