LIF388583874 Life Chemicals Inc.

{[5-(2-Thienyl)isoxazol-3-yl]methyl}amine hydrochloride

  • MDL number MFCD18483360
  • Cas Number : 1323536-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart