LIF388583876 Life Chemicals Inc.

{[5-(4-Methylphenyl)isoxazol-3-yl]methyl}amine hydrochloride

  • MDL number MFCD11519410
  • Cas Number : 1323584-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart