LIF388583881 Life Chemicals Inc.

4-(2-Fluorophenyl)-3-methyl-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD11506626
  • Cas Number : 1257878-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart