LIF388583882 Life Chemicals Inc.

8-oxa-3-azabicyclo[3.2.1]octane

  • MDL number MFCD09836264
  • Cas Number : 280-13-7
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart