LIF388583883 Life Chemicals Inc.

1-Benzyl-4-azepanone hydrochloride

  • MDL number MFCD11053515
  • Cas Number : 1208-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart