LIF388583886 Life Chemicals Inc.

4-(1-Piperazinyl)-1H-pyrazolo[3,4-d]pyrimidine dihydrochloride

  • MDL number MFCD22587895
  • Cas Number : 853680-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart