LIF388583887 Life Chemicals Inc.

Methyl 6-amino-1H-indole-4-carboxylate hydrochloride

  • MDL number MFCD09258897
  • Cas Number : 731810-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart