LIF388583888 Life Chemicals Inc.

4-(2-hydrazinoethyl)morpholine dioxalate

  • MDL number MFCD12922971
  • Cas Number : 1182284-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart