LIF388583895 Life Chemicals Inc.

{2-[(4-methyl-4H-1,2,4-triazol-3-yl)thio]ethyl}amine dihydrobromide

  • MDL number MFCD16660419
  • Cas Number : 1351609-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart