LIF388583896 Life Chemicals Inc.

3-amino-6-bromopyridin-2-ol hydrobromide

  • MDL number MFCD09701347
  • Cas Number : 1187930-34-2
  • Molecular Weight : 269.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart