LIF388583898 Life Chemicals Inc.

({3-[(4-methoxyphenoxy)methyl]-1,2,4-oxadiazol-5-yl}methyl)amine hydrochloride

  • MDL number MFCD16660415
  • Cas Number : 1245568-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart