LIF388583900 Life Chemicals Inc.

3-(4-methoxyphenyl)pyrrolidine oxalate

  • MDL number MFCD04117733
  • Cas Number : 96877-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart