LIF388583902 Life Chemicals Inc.

2-[4-(trifluoromethyl)phenyl]morpholine oxalate

  • MDL number MFCD03840015
  • Cas Number : 1198416-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart