LIF388583903 Life Chemicals Inc.

2-phenylmorpholine hydrochloride

  • MDL number MFCD03869024
  • Cas Number : 23972-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart