LIF388583904 Life Chemicals Inc.

3-[4-(ethoxycarbonyl)piperidin-1-yl]propanoic acid hydrochloride

  • MDL number MFCD22587932
  • Cas Number : 1351660-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart