LIF388583905 Life Chemicals Inc.

1-(cyclopropylsulfonyl)piperazine hydrochloride

  • MDL number MFCD16653430
  • Cas Number : 1057385-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart