LIF388583908 Life Chemicals Inc.

but-3-yn-2-amine hydrochloride

  • MDL number MFCD08752577
  • Cas Number : 42105-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart