LIF388583911 Life Chemicals Inc.

2-(2-piperazin-1-ylethyl)-1H-isoindole-1,3(2H)-dione dihydrochloride

  • MDL number MFCD08582805
  • Cas Number : 93897-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart