LIF388583912 Life Chemicals Inc.

8-hydrazinoquinoline dihydrochloride

  • MDL number MFCD00491379
  • Cas Number : 91004-61-4
  • Molecular Weight : 232.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart