LIF388583913 Life Chemicals Inc.

3-(2-aminoethyl)-7-methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one hydrochloride

  • MDL number MFCD22587939
  • Cas Number : 1351609-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart