LIF388583916 Life Chemicals Inc.

{2-[(4-methylphenyl)sulfonyl]ethyl}amine hydrochloride

  • MDL number MFCD03840167
  • Cas Number : 1093405-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart