LIF388583920 Life Chemicals Inc.

2-pyrrolidin-2-yl-1,3-benzothiazole hydrochloride

  • MDL number MFCD04966888
  • Cas Number : 1027643-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart