LIF388583930 Life Chemicals Inc.

(2-methylcyclopentyl)methanamine hydrochloride

  • MDL number MFCD16661139
  • Cas Number : 212382-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart