LIF388583931 Life Chemicals Inc.

6-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD08437638
  • Cas Number : 33537-97-2
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart