LIF394347880 Life Chemicals Inc.

2-methylimidazo[1,2-a]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD24387573
  • Cas Number : 1448074-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart