LIF398697738 Life Chemicals Inc.

[2-(2-methyl-1H-imidazol-1-yl)ethyl]amine dihydrobromide

  • MDL number MFCD26792904
  • Cas Number : 1156504-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart