LIF432341197 Life Chemicals Inc.

4-(3,4-Dichloro-phenyl)-thiazol-2-ylamine

  • MDL number MFCD00466323
  • Cas Number : 39893-80-6
  • Molecular Weight : 245.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart