LIF432341204 Life Chemicals Inc.

1-[2-amino-4-(thiophen-2-yl)-1,3-thiazol-5-yl]-2,2,2-trifluoroethan-1-one

  • MDL number MFCD01464537
  • Cas Number : 306289-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart