LIF432341205 Life Chemicals Inc.

3-(2-aminothiazol-4-yl)-2H-chromen-2-one

  • MDL number MFCD00481614
  • Cas Number : 61636-28-0
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart