LIF432341212 Life Chemicals Inc.

4,6-dimethylpyrimidine-2-thiol hydrochloride

  • MDL number MFCD00035317
  • Cas Number : 62501-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart