LIF432341215 Life Chemicals Inc.

8-formyl-4H-benzo[d][1,3]dioxine-6-carboxylic acid

  • MDL number MFCD01235547
  • Cas Number : 170807-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart