LIF432341224 Life Chemicals Inc.

4-(5-(4-bromophenyl)-3-(4-methoxyphenyl)-4,5-dihydro-1H-pyrazol-1-yl)-4-oxobutanoic acid

  • MDL number MFCD01152766
  • Cas Number : 309925-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart