LIF432341227 Life Chemicals Inc.

1-Acetyl-1,2-dihydro-indol-3-one

  • MDL number MFCD00466593
  • Cas Number : 16800-68-3
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart