LIF432341249 Life Chemicals Inc.

8-bromo-3-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD00462528
  • Cas Number : 93703-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart