LIF432341259 Life Chemicals Inc.

3-methyl-1-(4-nitrophenyl)piperazine hydrochloride

  • MDL number MFCD00545967
  • Cas Number : 1158439-97-4
  • Molecular Weight : 257.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart