LIF432341286 Life Chemicals Inc.

2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxamide

  • MDL number MFCD00687506
  • Cas Number : 24248-69-9
  • Molecular Weight : 211.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart