LIF432341289 Life Chemicals Inc.

4-(thiophen-2-yl)thiazol-2-amine

  • MDL number MFCD00457532
  • Cas Number : 28989-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart