LIF432341292 Life Chemicals Inc.

ethyl 3-amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD00124672
  • Cas Number : 52505-56-3
  • Molecular Weight : 250.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart