LIF432341309 Life Chemicals Inc.

methyl 2-hydroxyquinoline-4-carboxylate

  • MDL number MFCD01013783
  • Cas Number : 39497-01-3
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart