LIF432341310 Life Chemicals Inc.

methyl 2-chloroquinoline-4-carboxylate

  • MDL number MFCD03413689
  • Cas Number : 62482-26-2
  • Molecular Weight : 221.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart