LIF432341318 Life Chemicals Inc.

5-bromo-1H,3H-benzo[de]isochromene-1,3-dione

  • MDL number MFCD00228162
  • Cas Number : 24050-49-5
  • Molecular Weight : 277.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart