LIF432341331 Life Chemicals Inc.

2-methyl-4-(2-(4-phenylcyclohexylidene)hydrazinyl)quinoline hydrochloride

  • MDL number MFCD22586715
  • Cas Number : 1421497-63-3
  • Molecular Weight : 365.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart