LIF432341364 Life Chemicals Inc.

((4-(o-tolyloxy)phenyl)sulfonyl)leucine

  • MDL number MFCD03619075
  • Cas Number : 1008956-66-8
  • Molecular Weight : 377.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart